741507
1,6-Dihydro-1-methyl-6-oxo-3-pyridinecarboxylic acid
97%
Synonym(s):
Nudifloric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
mp
240-245 °C
functional group
carboxylic acid
storage temp.
2-8°C
SMILES string
CN1C=C(C=CC1=O)C(O)=O
InChI
1S/C7H7NO3/c1-8-4-5(7(10)11)2-3-6(8)9/h2-4H,1H3,(H,10,11)
InChI key
RGZCKPXTNJAWMR-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Urinary excretion of N1-methyl-2-pyridone-5-carboxylic acid and the fate of remaining of trigonelline.
Advances in experimental medicine and biology, 398, 599-603 (1996-01-01)
Journal of pharmaceutical and biomedical analysis, 30(3), 773-780 (2002-10-09)
Caffeine metabolites were extracted from urine samples collected 4 h after consumption of a cup of coffee and were separated by high-performance liquid chromatography (HPLC) on a C18 (5 microm) reverse-phase column using an acetonitrile (5%), acetic acid (0.05%) solution
The excretion of N1-methyl-2-pyridone-5-carboxylic acid and related compounds in human subjects after oral administration of nicotinic acid, trigonelline and N1-methyl-2-pyridone-5-carboxylic acid.
Advances in experimental medicine and biology, 294, 475-479 (1991-01-01)
Nicotinamide and insulin sensitivity.
Diabetologia, 36(4), 369-369 (1993-04-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service