Skip to Content
Merck
All Photos(1)

Key Documents

740551

Sigma-Aldrich

1α,25-Dihydroxyvitamin D3 (6,19,19-d3) solution

5 μg/mL in ethanol, 97 atom % D, 96% (CP)

Synonym(s):

1α,25-Dihydroxycholecalciferol (6,19,19-d3) solution, Calcitriol (6,19,19-d3) solution, Deuterated 1-α25-Dihydroxyvitamin D3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D3H41O3
CAS Number:
Molecular Weight:
419.65
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

97 atom % D

Quality Level

Assay

96% (CP)

form

solution

concentration

5 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

mp

-144.0 °C

application(s)

detection

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1[C@@H](O)C[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1/i2D2,11D

InChI key

GMRQFYUYWCNGIN-POLIPXRRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1α,25-Dihydroxyvitamin D3 [1,25(OH)2D3] is also known as calcitriol. It is known to monitor calcium and phosphate metabolism in humans. It also regulates insulin secretion and induces cellular differentiation.
1α,25-dihydroxyvitamin D3 (6,19,19-d3) is a deuterated vitamin wherein C-6 and C-19 protons are replaced by deuterium.

Application

1α,25-Dihydroxyvitamin D3 (6,19,19-d3) can be used as an analytical standard for accurate data interpretation in specific biological samples by mass spectrometry (MS).

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The emerging role of vitamin D3 in women?s health
Khadilkar, SS
Journal of Obstetrics and Gynaecology of India, 63(2), 147?150-147?150 (2013)
Janaina S Martins et al.
Endocrinology, 160(10), 2204-2214 (2019-06-27)
Phosphate homeostasis is critical for many cellular processes and is tightly regulated. The sodium-dependent phosphate cotransporter, NaPi2a, is the major regulator of urinary phosphate reabsorption in the renal proximal tubule. Its activity is dependent upon its brush border localization that
Vitamin D3: a helpful immuno-modulator
Di R, et al.
Immunology, 134(2), 123-139 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service