Skip to Content
Merck
All Photos(1)

Key Documents

H-089

Supelco

1α,25-Dihydroxyvitamin D3 solution

5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

5 μg/mL in ethanol

application(s)

clinical testing
clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

CC(CCCC(C)(C)O)C1CCC2C(\CCCC12C)=C\C=C3\CC(O)CC(O)C3=C

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-

InChI key

GMRQFYUYWCNGIN-KWJBEKLWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1α,25-Dihydroxyvitamin D3 is a biologically active form of vitamin D3 in calcium absorption and deposition. It has widespread effects on cellular differentiation and proliferation, and can modulate immune responsiveness, and central nervous system function. Recent studies suggest that 1α,25-dihydroxyvitamin D3 acts as a chemopreventive agent against several malignancies including cancers of the prostate and colon and shows synergy with other anticancer compounds.
A Certified Spiking Solution® suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls and calibrators for LC/UV and LC-MS/MS Vitamin D assays; as system tuning/suitability standards; linearity verification; internal proficiency testing; and for OEM use in preparation of reagents, development of assays and instrument IQ/OQ. The stable, quantitative solution format provides significant benefits to the user including material savings, increased efficiency of labor, and convenience of use. The shelf stable Snap-N-Spike® format allows laboratories to eliminate painstaking weighing operations and provides the ability to spike a known concentration of analyte into their matrix of choice just before use.

Application

1α,25-Dihydroxyvitamin D3 may be used as a certified reference material for the determination of the analyte in biological fluids by liquid chromatography-tandem mass spectrometry.[1][2][3]

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 1, 25-dihydroxyvitamin D2 and 1, 25-dihydroxyvitamin D3 in human serum using liquid chromatography with tandem mass spectrometry
Fang H, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1027, 19-26 (2016)
Sensitive analysis of 1?, 25-dihydroxyvitamin D3 in biological fluids by liquid chromatography--tandem mass spectrometry
Kissmeyer AM and Sonne K
Journal of Chromatography A, 935(1-2), 93-103 (2001)
Zhican Wang et al.
Analytical biochemistry, 418(1), 126-133 (2011-07-26)
Simultaneous and accurate measurement of circulating vitamin D metabolites is critical to studies of the metabolic regulation of vitamin D and its impact on health and disease. To that end, we have developed a specific liquid chromatography-tandem mass spectrometry (LC-MS/MS)
R Volchenkov et al.
Scandinavian journal of immunology, 77(4), 246-254 (2013-02-28)
Dendritic cells (DC) are professional antigen-presenting cells that are capable of both activating immune responses and inducing tolerance. Several studies have revealed efficiency of therapeutic vaccination with tolerogenic DC (tolDC) in inhibition of experimental autoimmunity. The purpose of this study
Nicola Napoli et al.
PloS one, 8(2), e56488-e56488 (2013-02-26)
Vitamin D supplementation in childhood improves the achievement of peak bone mass. We investigated the effect of supplementation with calcitriol on bone turnover in recent-onset type 1 diabetes (T1D). Moreover, the association between osteocalcin and parameters of β-cell function and

Articles

Novel sample prep technique coupled with the unique selectivity of the Ascentis Express F5 column enables a fast and simplified bioanalytical method for associated vitamin D metabolites.

Using HybridSPE-Phospholipid, Ascentis Express, and Other LC-MS Workflow Components

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service