Skip to Content
Merck
All Photos(1)

Key Documents

740233

Sigma-Aldrich

25-Hydroxyvitamin D2 solution

5 μg/mL in ethanol, 98% (CP)

Synonym(s):

25-Hydroxycalciferol solution, 25-Hydroxyergocalciferol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O2
Molecular Weight:
412.65
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

98% (CP)

form

solution

concentration

5 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

storage temp.

−20°C

SMILES string

C[C@H](\C=C\[C@H](C)C(C)(C)O)[C@H]1CC[C@H]2C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C28H44O2/c1-19-10-14-24(29)18-23(19)13-12-22-8-7-17-28(6)25(15-16-26(22)28)20(2)9-11-21(3)27(4,5)30/h9,11-13,20-21,24-26,29-30H,1,7-8,10,14-18H2,2-6H3/b11-9+,22-12+,23-13-/t20-,21+,24+,25-,26+,28-/m1/s1

InChI key

KJKIIUAXZGLUND-ICCVIKJNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

25-Hydroxyvitamin D2 (ergocalciferol or 25-hydroxycalciferol) is a metabolite of vitamin D found in the liver. It is one of the most common forms of vitamin D circulating in the body and plays an important role in determining vitamin D deficiency.

Application

25-Hydroxyvitamin D2 can be used as an analytical standard for the quantitative analysis of 25-Hydroxyvitamin D2 content in biological fluids via LC-MS/MS.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determinants of vitamin D deficiency from Sun exposure: a global perspective
Vitamin D, 19(1), 79-90 (2018)
Maarten W Taal et al.
Kidney international, 86(2), 407-413 (2014-01-17)
Fibroblast growth factor (FGF) 23 is an important regulator of phosphaturia. Its serum level was found to increase before that of parathyroid hormone (PTH) in early chronic kidney disease (CKD) in some but not all previous studies. As vitamin D
10 years of 25-hydroxyvitamin-D testing by LC-MS/MS-trends in vitamin-D deficiency and sufficiency
Galior K, et al.
Bone Reports, 8(1), 268-273 (2018)
Development and validation of an LC-MS/MS based method for quantification of 25 hydroxyvitamin D2 and 25 hydroxyvitamin D3 in human serum and plasma
Zhang SW, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 961(1), 62-70 (2014)
Vitamin-D2 treatment-associated decrease in 25 (OH) D3 level is a reciprocal phenomenon: a randomized controlled trial
Hammami, MM et al.
BMC endocrine disorders, 19(1), 8-8 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service