Skip to Content
Merck
All Photos(1)

Key Documents

739669

Sigma-Aldrich

25-Hydroxyvitamin D3 solution

5 μg/mL in ethanol, 98% (CP)

Synonym(s):

25-Hydroxycholecalciferol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

98% (CP)

form

solution

concentration

5 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

storage temp.

−20°C

SMILES string

C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-DTOXIADCSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

25-Hydroxyvitamin D3 (calcifediol) is a prohormone produced via hydroxylation of vitamin D3 (cholecalciferol) in the liver. It is a precursor for the synthesis of calcitriol {1,25-dihydroxyvitamin D3 or [1,25(OH)2D3]}. It is also used as a biomarker to determine the status of vitamin D in the body.

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina V Antunes et al.
Therapeutic drug monitoring, 37(6), 733-744 (2015-04-09)
In view of the large variability on therapeutic response and the multiple factors associated to tamoxifen (TAM) metabolic activation, this study aimed to evaluate the effect of CYP2D6 and CYP3A4 phenotypes, drug interactions, and vitamin D exposure on TAM metabolism
Peter Daley et al.
The Lancet. Infectious diseases, 15(5), 528-534 (2015-04-13)
Vitamin D has immunomodulatory effects that might aid clearance of mycobacterial infection. We aimed to assess whether vitamin D supplementation would reduce time to sputum culture conversion in patients with active tuberculosis. We did this randomised, double-blind, placebo-controlled, superiority trial
Alberto Lo Gullo et al.
PloS one, 10(8), e0134602-e0134602 (2015-08-05)
Suboptimal vitamin D status was recently acknowledged as an independent predictor of cardiovascular diseases and all-cause mortality in several clinical settings, and its serum levels are commonly reduced in Rheumatoid Arthritis (RA). Patients affected by RA present accelerated atherosclerosis and
Fadia S AlBuhairan et al.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine, 57(3), 263-269 (2015-08-25)
With the increasing burden of noncommunicable disease, adolescence is viewed as an opportune time to prevent the onset of certain behaviors and promote healthy states. Although adolescents comprise a considerable portion of Saudi Arabia's population, they have received insufficient attention
Joseph Schwager et al.
Molecular nutrition & food research, 59(8), 1513-1523 (2015-03-27)
The impact of L-ascorbic acid (L-AA) on the chemokinesis (CK) and chemotaxis (CT) of HL-60 cells and polymorphonuclear cells (PMN) was investigated. HL-60 cells were differentiated with DMSO, retinoic acid (RA), vitamin D, or L-AA. Chemokinesis and chemotaxis of differentiated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service