Skip to Content
Merck
All Photos(2)

Key Documents

734527

Sigma-Aldrich

Tetrakis(pyridine)copper(II) triflate

95%

Synonym(s):

[Cu(OTf)2(py)4], Cu(pyridine)4(OTf)2, Tetrakis(pyridine)copper(II) bis(trifluoromethanesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20CuF6N4O6S2
CAS Number:
Molecular Weight:
678.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

crystals

reaction suitability

core: copper
reagent type: catalyst

mp

190-194 °C

storage temp.

−20°C

SMILES string

c1ccncc1.c2ccncc2.c3ccncc3.c4ccncc4.FC(F)(F)S(=O)(=O)O[Cu]OS(=O)(=O)C(F)(F)F

InChI

1S/4C5H5N.2CHF3O3S.Cu/c4*1-2-4-6-5-3-1;2*2-1(3,4)8(5,6)7;/h4*1-5H;2*(H,5,6,7);/q;;;;;;+2/p-2

InChI key

DOUMBTAZXRQKGF-UHFFFAOYSA-L

Application

Tetrakis(pyridine)copper(II) triflate [Cu(OTf)2(py)4] is a general copper-based catalyst. It is commonly used to catalyze copper-mediated radiofluorination of arenes and heteroarene. It is also employed in C(sp3)-H functionalization/cyclisation of 2-amino-N,N-dialkylbenzamides to synthesize 2,3-disubstituted-4(3H)-quinazolinones.
Tetrakis(pyridine)copper(II) triflate can be used to catalyze:
  • Intra and intermolecular aziridination of allylic carbamates and styrenes.
  • 18F-fluorination of aryl boronic esters to generate fluroarenes applicable as radiotracers.
  • Nucleophilic radiobromination of aryl borons with a radiobromide ion applicable in the preparation of radiobrominated pharmaceuticals.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed alkene aziridination with N-tosyloxycarbamates
Lebel H, et al.
Organic Letters, 9(23), 4797-4800 (2007)
Preparation of No?Carrier?Added 6?[18F] Fluoro?l?tryptophan via Cu?Mediated Radiofluorination.
Schafer D, et al.
European Journal of Organic Chemistry, 2016(27), 4621-4628 (2016)
Thomas Keller et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 40(5), 1012-1020 (2019-05-31)
[18F]F-DPA, a novel translocator protein 18 kDa (TSPO)-specific radioligand for imaging neuroinflammation, has to date been synthesized with low to moderate molar activities (Am's). In certain cases, low Am can skew the estimation of specific binding. The high proportion of the
Pyrazine and pyridine complexes of copper (II) trifluoromethanesulfonate. Crystal structure of tetrakis (pyridine) bis (trifluoromethanesulfonato-O) copper (II) and magnetic exchange in (pyrazine) bis (trifluoromethanesulfonato-O) copper (II).
Haynes J S, et al.
Inorganic Chemistry, 27(7), 1237-1241 (1988)
Copper-catalyzed C?H functionalization reactions: Efficient synthesis of heterocycles.
Guo X X, et al.
Chemical Reviews, 115(3), 1622-1651 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service