Skip to Content
Merck
All Photos(3)

Key Documents

722219

Sigma-Aldrich

1-Benzyl-3-hydroxy-1H-indazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O
CAS Number:
Molecular Weight:
224.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

165-169 °C

SMILES string

Oc1nn(Cc2ccccc2)c3ccccc13

InChI

1S/C14H12N2O/c17-14-12-8-4-5-9-13(12)16(15-14)10-11-6-2-1-3-7-11/h1-9H,10H2,(H,15,17)

InChI key

SXPJFDSMKWLOAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of 1-benzyl-1H-indazol-3-ol in benzydamine in pharmaceutical formulations.
G Carlucci et al.
Journal of pharmaceutical and biomedical analysis, 14(5), 655-657 (1996-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service