Skip to Content
Merck
All Photos(1)

Key Documents

718165

Sigma-Aldrich

Trioctylphosphine

97%

Synonym(s):

TOP, P(Oct)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]3P
CAS Number:
Molecular Weight:
370.64
Beilstein:
1776995
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.468 (lit.)

bp

284-291 °C/50 mmHg (lit.)

density

0.831 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CCCCCCCCP(CCCCCCCC)CCCCCCCC

InChI

1S/C24H51P/c1-4-7-10-13-16-19-22-25(23-20-17-14-11-8-5-2)24-21-18-15-12-9-6-3/h4-24H2,1-3H3

InChI key

RMZAYIKUYWXQPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trioctylphosphine serves as a phosphorus source and used in the synthesis of metal nanoparticles.

Application

Trioctylphosphine can be used for the conversion of metal nanocrystals, bulk powders, foils, wires, thin films to metal phosphides. It can also act as a solvent and stabilizer for synthesizing cadmium sulfide nanorods from cadmium acetate and sulfur.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

296.6 °F - closed cup

Flash Point(C)

147 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trioctylphosphine as both solvent and stabilizer to synthesize CdS nanorods.
Chen S, et al.
Nanoscale Research Letters, 4(10), 1159-1159 (2009)
Palladium (ii)-complexes of bi-and tri-dentate phosphine ligands: precursor for palladium--phosphorous nanoparticles and activity towards Suzuki--Miyaura coupling
Rao GK, et al.
New. J. Chem., 47, 4218-4227 (2023)
Trioctylphosphine: a general phosphorus source for the low-temperature conversion of metals into metal phosphides.
Henkes AE & Schaak RE.
Chemistry of Materials, 19(17), 4234-4242 (2007)
Tilman Läppchen et al.
EJNMMI physics, 7(1), 22-22 (2020-04-24)
For multicenter clinical studies, PET/CT and SPECT/CT scanners need to be validated to ensure comparability between various scanner types and brands. This validation is usually performed using hollow phantoms filled with radioactive liquids. In recent years, 3D printing technology has
Kimberly H Hartstein et al.
ACS nano, 11(10), 10430-10438 (2017-09-16)
We examine the effects of CdS shell growth on photochemical reduction of colloidal CdSe quantum dots (QDs) and describe the spectroscopic properties of the resulting n-type CdSe/CdS QDs. CdS shell growth greatly slows electron trapping. Because of this improvement, complete

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service