Skip to Content
Merck
All Photos(1)

Key Documents

74740

Sigma-Aldrich

1-Octadecene

≥95.0% (GC)

Synonym(s):

α-Octadecene, AlphaPlus 1-Octadecene, Neodene 18

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CH=CH2
CAS Number:
Molecular Weight:
252.48
Beilstein:
1761067
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

0.71 (vs air)

Quality Level

Assay

≥95.0% (GC)

form

liquid

autoignition temp.

482 °F

refractive index

n20/D 1.444 (lit.)
n20/D 1.445

bp

314.4 +/- 5.0 °C/760 mmHg (lit.)
179 °C/15 mmHg (lit.)
314.4 °C±5.0 °C/760 mmHg (lit.)

mp

14-16 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

CCCCCCCCCCCCCCCCC=C

InChI

1S/C18H36/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3H,1,4-18H2,2H3

InChI key

CCCMONHAUSKTEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The copolymerization of ethylene and 1-octadecene using a bridged metallocene has been studied. Monolayers of 1-octadecene were attached on a hydrogen-terminated (111) silicon surface via free-radical reaction.

Application

1-Octadecene may be used in the preparation of monodisperse CuPd alloy nanoparticles (NPs). It may be used in the synthesis of magnetite nanocrystals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of copolymers of ethylene and 1-octadecene using the rac-Et (Ind) 2ZrCl2/MAO catalyst system.
Quijada R, et al.
Macromol. Chem. Rapid Commun., 200(6), 1306-1310 (1999)
1-octadecene monolayers on Si (111) hydrogen-terminated surfaces: Effect of substrate doping.
Miramond C and Vuillaume D.
Journal of Applied Physics, 96(3), 1529-1536 (2004)
The effect of nanocrystalline magnetite size on arsenic removal.
Mayo J T, et al.
Science and Technology of Advanced Materials, 8(1), 71-75 (2007)
Sümeyra Diyarbakir et al.
ACS applied materials & interfaces, 7(5), 3199-3206 (2015-01-17)
Monodisperse CuPd alloy nanoparticles (NPs) were prepared by using a typical high-temperature organic solution phase protocol comprising the coreduction of copper(II) acetylacetonate and palladium(II) acetylacetonate by morpholine-borane complex in oleylamine and 1-octadecene solution at 80 °C. The presented synthesis protocol
Oleksandr Voznyy et al.
ACS nano, 13(10), 11122-11128 (2019-09-21)
Colloidal quantum dots (CQDs) allow broad tuning of the bandgap across the visible and near-infrared spectral regions. Recent advances in applying CQDs in light sensing, photovoltaics, and light emission have heightened interest in achieving further synthetic improvements. In particular, improving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service