Skip to Content
Merck
All Photos(1)

Key Documents

714887

Sigma-Aldrich

2,2′-Azobis(2-methylpropionitrile) solution

0.2 M in toluene

Synonym(s):

α,α,′-Azoisobutyronitrile solution, AIBN solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4
CAS Number:
Molecular Weight:
164.21
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

0.2 M in toluene

refractive index

n20/D 1.495

density

0.858 g/mL at 25 °C

functional group

azo
nitrile

storage temp.

2-8°C

SMILES string

CC(C)(\N=N\C(C)(C)C#N)C#N

InChI

1S/C8H12N4/c1-7(2,5-9)11-12-8(3,4)6-10/h1-4H3/b12-11+

InChI key

OZAIFHULBGXAKX-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2,2′-Azobis(2-methylpropionitrile) (AIBN) is a radical initiator. AIBN solution can be used to initiate radical-induced reactions, specifically free-radical polymerizations. It can be used in:
  • Synthesis of styrene-vinyl pyridine diblock copolymers by reversible addition-fragmentation chain transfer (RAFT) polymerization.
  • Preparation of silicon oxycarbide glasses.
  • Synthesis of poly [N-(p-vinyl benzyl) phthalimide] for the preparation of titanium dioxide composites for electrophoretic displays.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Styrene?vinyl pyridine diblock copolymers: Synthesis by RAFT polymerization and self?assembly in solution and in the bulk.
Zamfir M, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 50(8), 1636-1644 (2012)
Stable titanium dioxide grafted with poly [N-(p-vinyl benzyl) phthalimide] composite particles in suspension for electrophoretic displays.
Dai R, et al.
Colloid and Polymer Science, 289(4), 401-407 (2011)
Cameron J Bloomquist et al.
Journal of controlled release : official journal of the Controlled Release Society, 278, 9-23 (2018-03-30)
Mass customization along with the ability to generate designs using medical imaging data makes 3D printing an attractive method for the fabrication of patient-tailored drug and medical devices. Herein we describe the application of Continuous Liquid Interface Production (CLIP) as
Optimization of sol?gel/pyrolysis routes to silicon oxycarbide glasses.
Strachota A, et al.
Journal of Non-Crystalline Solids, 358(20), 2771-2782 (2012)
Aspects of the chemistry of azobisisobutyronitrile.
Bevington J C.
Journal of Polymer Science, 10(3), 195-202 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service