Skip to Content
Merck
All Photos(1)

Key Documents

708011

Sigma-Aldrich

Amano Lipase PS

(immobilized on diatomite)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

form

powder

storage temp.

2-8°C

Application

Amano Lipase PS can be used as a catalyst in the enzymatic esterification reactions. It is also used in the resolution of racemic mixtures such as (R)- and (S)-1-trimethylsilyl-1-alkyn-3-ols by enantioselective acetylation reactions.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miniemulsion as efficient system for enzymatic synthesis of acid alkyl esters
De Barros D PC, et al.
Biotechnology and Bioengineering, 106(4), 507-515 (2010)
Lipase catalysed resolution of (R)-and (S)-1-trimethylsilyl-1-alkyn-3-ols: useful intermediates for the synthesis of optically active ?-lactones
Allevi P, et al.
Tetrahedron Asymmetry, 8(1), 93-99 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service