Skip to Content
Merck
All Photos(1)

Key Documents

248932

Sigma-Aldrich

(R)-(−)-1,1′-Binaphthyl-2,2′-diyl hydrogenphosphate

≥98%

Synonym(s):

(R)-(−)-1,1′-Binaphthalene-2,2′-diyl hydrogen phosphate, (R)-4-Hydroxydinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13O4P
CAS Number:
Molecular Weight:
348.29
Beilstein:
4713363
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

optical activity

[α]20/D −605°, c = 1.35 in methanol

functional group

phosphate

InChI

1S/C20H13O4P/c21-25(22)23-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)24-25/h1-12H,(H,21,22)

InChI key

JEHUZVBIUCAMRZ-UHFFFAOYSA-N

Application

(R)-(−)-1,1′-Binaphthyl-2,2′-diyl hydrogenphosphate is an organocatalyst that can be used:       
  • To facilitates the controlled ring-opening homopolymerization and copolymerization of ε-caprolactone (cyclic ester).        
  • As an additive along with a copper catalyst to achieve the highest catalytic activity in Sonogashira-type reactions.

A chiral ligand used in hydrocarboxylation reactions. Complexes with rhodium and mediates the asymmetric dipolar cycloaddition of diazo compounds. A number of racemic amines which have proven difficult to separate have been resolved with this chiral acid. Palladium derivatives have been used in asymmetric hydrocarboxylations, and rhodium derivatives have been used in dipolar cycloadditions.
Used in a chiral Bronsted acid catalyzed enantoselective Mannich reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organophosphate-accelerated copper-catalyzed C (sp2)-C (sp) Sonogashira-type cross couplings
Xu W, et al.
Applied Organometallic Chemistry, 29(5), 301-304 (2015)
Controlled ring-opening polymerization of cyclic esters with phosphoric acid as catalysts
Zhou Xi and Hong L
Colloid and Polymer Science, 291(9), 2155-2162 (2013)
Tetrahedron Letters, 33, 5987-5987 (1992)
Journal of the American Chemical Society, 112, 2803-2803 (1990)
Craig, J.C. Ansari, A.M.
Chirality, 5, 188-188 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service