Skip to Content
Merck
All Photos(1)

Key Documents

703524

Sigma-Aldrich

Lithium tert-amoxide solution

40 % (w/w) in heptanes

Synonym(s):

Lithium 2-methyl-2-butoxide, Lithium tert-amylate, Lithium tert-pentoxide, Lithium tert-pentyloxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11LiO
CAS Number:
Molecular Weight:
94.08
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

40 % (w/w) in heptanes
~3.1 M in heptane

density

0.73 g/mL at 25 °C

SMILES string

[Li+].CCC(C)(C)[O-]

InChI

1S/C5H11O.Li/c1-4-5(2,3)6;/h4H2,1-3H3;/q-1;+1

InChI key

QPDJILZPDAMLFH-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

15.1 °F

Flash Point(C)

-9.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service