Skip to Content
Merck
All Photos(3)

Key Documents

361798

Sigma-Aldrich

Lithium diisopropylamide solution

2.0 M in THF/heptane/ethylbenzene

Synonym(s):

LDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NLi
CAS Number:
Molecular Weight:
107.12
Beilstein:
3655042
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

2.0 M in THF/heptane/ethylbenzene

impurities

3 wt. % diisopropylamine

density

0.812 g/mL at 25 °C

functional group

amine

storage temp.

2-8°C

SMILES string

[Li]N(C(C)C)C(C)C

InChI

1S/C6H14N.Li/c1-5(2)7-6(3)4;/h5-6H,1-4H3;/q-1;+1

InChI key

ZCSHNCUQKCANBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium diisopropylamide solution (LDA) is a sterically hindered non-nucleophilic strong base widely used in organic synthesis.

Application

Lithium diisopropylamide solution (LDA) can be used:
  • As an initiator in the anionic polymerization of D,L-lactide and methyl methacrylate.
  • To facilitate ester enolization.
  • To convert carboxylic acids to enediolate intermediates for preparing trifluoromethyl ketones.
  • In ortho-lithiation of arylsulfonyloxazolidinones to prepare N-substituted saccharin analogs.
  • To catalyze ortholithiation and Fries rearrangement of aryl carbamates.
  • As a promoter in the isomerization of allylic ethers to (Z)-propenyl ethers.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, hearing organs, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium diisopropylamide-mediated enolizations: Solvent-dependent mixed aggregation effects
Sun X and Collum DB
Journal of the American Chemical Society, 122(11), 2459-2463 (2000)
Efficient access to chiral N-substituted saccharin analogues via the directed ortho-lithiation of 3-N-arylsulfonyloxazolidin-2-ones
Aliyenne AO, et al.
Tetrahedron Letters, 47(36), 6405-6408 (2006)
Trifluoromethyl ketones: properties, preparation, and application
Kelly CB, et al.
Chemical Communications (Cambridge, England), 49(95), 11133-11148 (2013)
Isomerization of allyl ethers initiated by lithium diisopropylamide
Su C, and Williard PG
Organic Letters, 12(23), 5378-5381 (2010)
Tetrahedron Letters, 47, 6405-6405 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service