Skip to Content
Merck
All Photos(3)

Key Documents

700207

Sigma-Aldrich

Polyetherimide

melt index 18 g/10 min (337 °C/6.6kg)

Synonym(s):

Poly(bisphenol A-co-4-nitrophthalic anhydride-co-1,3-phenylenediamine), PEI, Poly(bisphenol A anhydride-co-1,3-phenylenediamine), Poly(bisphenol A dianhydride-co-1,3-phenylenediamine), Poly(ether imide), Poly-[2,2′bis(4-(3,4-dicarboxyphenoxy)phenylpropane)-1,3-phenylene bisimide], Polyetherimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C39H30N2O6)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

pellets

Quality Level

melt index

18 g/10 min (337 °C/6.6kg)

density

1.27 g/mL at 25 °C (lit.)

SMILES string

Nc7cc(ccc7)N.O1C(=O)c2c(ccc(c2)Oc3ccc(cc3)C(C)(C)c4ccc(cc4)Oc5cc6c(cc5)C(=O)OC6=O)C1=O

InChI

1S/C31H20O8.C6H8N2/c1-31(2,17-3-7-19(8-4-17)36-21-11-13-23-25(15-21)29(34)38-27(23)32)18-5-9-20(10-6-18)37-22-12-14-24-26(16-22)30(35)39-28(24)33;7-5-2-1-3-6(8)4-5/h3-16H,1-2H3;1-4H,7-8H2

InChI key

VYLFLJXGGIUQKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assad U Khan et al.
Advanced materials (Deerfield Beach, Fla.), 33(2), e2004053-e2004053 (2020-11-26)
Graphene holds promise for thin, ultralightweight, and high-performance nanoelectromechanical transducers. However, graphene-only devices are limited in size due to fatigue and fracture of suspended graphene membranes. Here, a lightweight, flexible, transparent, and conductive bilayer composite of polyetherimide and single-layer graphene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service