686271
(S)-4-Chloro-α-methylbenzyl alcohol
95%
Synonym(s):
(S)-1-(4-Chlorophenyl)ethanol, (S)-4′-Chloro-1-phenylethanol
About This Item
Recommended Products
Quality Level
Assay
95%
form
liquid
optical activity
[α]20/D -48.0°, c = 1 in chloroform
refractive index
n20/D 1.544
density
1.175 g/mL at 25 °C
functional group
chloro
hydroxyl
SMILES string
C[C@H](O)c1ccc(Cl)cc1
InChI
1S/C8H9ClO/c1-6(10)7-2-4-8(9)5-3-7/h2-6,10H,1H3/t6-/m0/s1
InChI key
MVOSNPUNXINWAD-LURJTMIESA-N
Related Categories
Application
- The enantioselective synthesis of (-)-(S,S)-clemastine.
- The enantioselective and geometrically defined synthesis of chloro methylbenzyl pinacol boronic ester via lithiation−borylation methodology.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service