Skip to Content
Merck
All Photos(1)

Key Documents

147559

Sigma-Aldrich

1-(4-Chlorophenyl)ethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(CH3)OH
CAS Number:
Molecular Weight:
156.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

119 °C/10 mmHg (lit.)

density

1.171 g/mL at 25 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

CC(O)c1ccc(Cl)cc1

InChI

1S/C8H9ClO/c1-6(10)7-2-4-8(9)5-3-7/h2-6,10H,1H3

InChI key

MVOSNPUNXINWAD-UHFFFAOYSA-N

Application

1-(4-Chlorophenyl)ethanol was used to study transfer dehydrogenation of various alcohols over heterogeneous palladium catalysts using olefins as hydrogen acceptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective transfer dehydrogenation of aromatic alcohols on supported palladium.
Keresszegi C, et al.
New. J. Chem., 25(9), 1163-1167 (2001)
Xiao-Hong Chen et al.
BMC biotechnology, 11, 110-110 (2011-11-22)
Chiral alcohols are widely used in the synthesis of chiral pharmaceuticals, flavors and functional materials and appropriate whole-cell biocatalysts offer a highly enantioselective, minimally polluting route to these valuable compounds. The recently isolated strain Acetobacter sp. CCTCC M209061 showed exclusive
Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service