Skip to Content
Merck
All Photos(1)

Key Documents

680133

Sigma-Aldrich

2-Chloro-6-methylbenzaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO
CAS Number:
Molecular Weight:
154.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

bp

74 °C/0.4 mmHg

mp

36-40 °C

SMILES string

[H]C(=O)c1c(C)cccc1Cl

InChI

1S/C8H7ClO/c1-6-3-2-4-8(9)7(6)5-10/h2-5H,1H3

InChI key

CCYFXIJPJFSTSU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.0 °F

Flash Point(C)

98.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service