Skip to Content
Merck
All Photos(2)

Key Documents

674753

Sigma-Aldrich

Aluminum acetylacetonate

purified by sublimation, 99.999% trace metals basis

Synonym(s):

Al(acac)3, Aluminum 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(C5H7O2)3
CAS Number:
Molecular Weight:
324.31
Beilstein:
4157942
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

powder
solid

purified by

sublimation

reaction suitability

core: aluminum

bp

315 °C (lit.)

mp

190-193 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Al](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Al/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

KILURZWTCGSYRE-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

244.4 °F

Flash Point(C)

118 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Zatta et al.
Toxicology letters, 45(1), 15-21 (1989-01-01)
Tris acetylacetonate aluminium(III) (Al(acac)3), dissolved in water, is effective in producing osmotic fragility in suspended erythrocytes in the concentration range of 0.034-0.34 mmol/l. Water solutions of Tris maltolate aluminium(III) (Al(malt)3) and aluminium lactate (Al(lac)3) are also effective but the dose-response
G G Bombi et al.
Environmental health perspectives, 89, 217-223 (1990-11-01)
Aluminum lactate [Al(lact)3] (hydrophilic, hydrolytically unstable) and aluminum acetylacetonate [Al(acae)3] (lipophilic, hydrolytically stable) were tested as potential toxicants to rabbits upon IV administration both as aqueous solutions and as liposome suspensions. Both chemicals behaved as cardiotoxic agents when administered as
P Zatta et al.
Journal of inorganic biochemistry, 65(2), 109-114 (1997-02-01)
The action of aluminum [Al(III)] as Al(acac)3 on erythrocytes causes biophysical effects such as osmotic fragility and echino-acanthocytes formation. In this paper, we present these effects in terms of variation of membrane fluidity, together with findings regarding conformational modifications of
John W Logan et al.
Solid state nuclear magnetic resonance, 22(2-3), 97-109 (2002-12-10)
Additional experimental evidence of rotary resonance effects for multiple-quantum coherence conversion in a spin-5/2 system is presented. Two-dimensional plots of the relative efficiency of MQ excitation and conversion are given as a function of radio frequency (rf) amplitude and pulse
Low-temperature synthesis of gamma-alumina nanocrystals from aluminum acetylacetonate in nonaqueous media.
Shuxue Zhou et al.
Small (Weinheim an der Bergstrasse, Germany), 3(5), 763-767 (2007-04-07)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service