Skip to Content
Merck
All Photos(1)

Key Documents

669997

Sigma-Aldrich

Δ-TRISPHAT tetrabutylammonium salt

≥98.5% (31P-NMR)

Synonym(s):

[Tetrabutylammonium] [Δ-tris(tetrachloro-1,2-benzenediolato)phosphate(V)]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18Cl12O6P · C16H36N
CAS Number:
Molecular Weight:
1011.06
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.5% (31P-NMR)

form

solid

optical activity

[α]/D -370±10°, c = 0.1 in ethanol

storage temp.

2-8°C

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.Clc1c(Cl)c(Cl)c2O[P-]345(Oc2c1Cl)Oc6c(Cl)c(Cl)c(Cl)c(Cl)c6O3.Clc7c(Cl)c(Cl)c(O4)c(O5)c7Cl

InChI

1S/C18Cl12O6P.C16H36N/c19-1-2(20)8(26)14-13(7(1)25)31-37(32-14,33-15-9(27)3(21)4(22)10(28)16(15)34-37)35-17-11(29)5(23)6(24)12(30)18(17)36-37;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h;5-16H2,1-4H3/q-1;+1

InChI key

GCGVBYHPNKLLAV-UHFFFAOYSA-N

General description

Δ-TRISPHAT tetrabutylammonium salt, containing a hexacoordinated phosphorus anion, is a chiral NMR solvating and asymmetry-inducing reagent.

Application

Chiral anion mediated asymmetric chemistry

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C3-Symmetric Trinuclear Molybdenum Cluster Sulfides: Configurational Stability, Supramolecular Stereocontrol, and Absolute Configuration Assignment.
Frantz R, et al.
Inorganic Chemistry, 46(25), 10717-10723 (2007)
Application of TRISPHAT anion as NMR chiral shift reagent.
Lacour Je, et al.
Chemical Communications (Cambridge, England), 23, 2285-2286 (1997)
Jerome Lacour et al.
Organic & biomolecular chemistry, 3(1), 15-19 (2004-12-17)
Chemical reactions and processes often involve chiral, yet racemic, cationic reagents, intermediates or products. To afford instead non racemic or enantiopure compounds, an asymmetric ion pairing of the cations with enantiopure anions can be considered--the counter ions behaving as asymmetric
Jérôme Lacour et al.
Chemical Society reviews, 32(6), 373-382 (2003-12-16)
Chemical reactions and processes often involve cationic prostereogenic or racemic reagents, intermediates or products. To afford instead non-racemic or enantiopure compounds, an asymmetric ion pairing of the cations with chiral anionic counterions can be considered. This review presents recent examples
Bimacrocyclic concave N-heterocyclic carbenes (NHCs): Synthesis, structure and application in catalyses.
Winkelmann O and Luning U.
Supramolecular Chemistry, 21(3-4), 223-229 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service