Skip to Content
Merck
All Photos(1)

Key Documents

668796

Sigma-Aldrich

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine

97%

Synonym(s):

(S)-(+)-1-AminoTetralin, (S)-(+)-1-Aminotetraline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H11)NH2
CAS Number:
Molecular Weight:
147.22
Beilstein:
2360279
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.565

bp

250 °C

density

1.010 g/mL at 25 °C

SMILES string

N[C@H]1CCCc2ccccc12

InChI

1S/C10H13N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7,11H2/t10-/m0/s1

InChI key

JRZGPXSSNPTNMA-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is a chiral amine that can be used:
  • To prepare a 2-pyridylmethylene-(S)-(+)-1,2,3,4-tetrahydro-1-naphthylamine containing pentanuclear trigonal bipyramidal [Co3Fe2] complex, which exhibits electron-transfer-coupled spin transition (ETCST) properties.
  • To prepare imine-substituted thiophene Schiff base compounds.
  • To derivatize macrocyclic diarylheptanoid compounds of biological importance.

Legal Information

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and antibacterial evaluation of macrocyclic diarylheptanoid derivatives
Lin H, et al.
Bioorganic & medicinal chemistry letters, 26(16), 4070-4076 (2016)
Solvent-induced on/off switching of intramolecular electron transfer in a cyanide-bridged trigonal bipyramidal complex
Wei R-J, et al.
Dalton Transactions, 45(43), 17104-17107 (2016)
Crystal structures of four chiral imine-substituted thiophene derivatives
Hern'andez-T'ellez G, et al.
Acta Crystallographica Section E: Crystallographic Communications, 72(3), 350-354 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service