Skip to Content
Merck
All Photos(1)

Key Documents

662682

Sigma-Aldrich

Cytidine-15N3 5′-monophosphate disodium salt

≥98 atom % 15N, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H1215N3Na2O8P
Molecular Weight:
370.14
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % 15N

Quality Level

Assay

≥95% (CP)

form

solid

technique(s)

bio NMR: suitable

mass shift

M+3

SMILES string

OC1C(COP(O[Na])(O[Na])=O)OC([15N]2C([15N]=C([15NH2])CC2)=O)C1O

InChI

1S/C9H14N3O8P.2Na/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1/i10+1,11+1,12+1;;

InChI key

INTPYBRGLGSMRA-RNONBHISSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Molecular weight and formula reflect free acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service