Skip to Content
Merck
All Photos(1)

Key Documents

662666

Sigma-Aldrich

Uridine-15N2 5′-monophosphate disodium salt

≥98 atom % 15N, ≥95% (CP)

Synonym(s):

5′-Uridylic-15N2 acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H1115N2Na2O9P
Molecular Weight:
370.13
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % 15N

Quality Level

Assay

≥95% (CP)

form

solid

technique(s)

bio NMR: suitable

mass shift

M+2

storage temp.

−20°C

SMILES string

OC1C(COP(O[Na])(O[Na])=O)OC([15N]2C=CC([15N]([H])C2=O)=O)C1O

InChI

1S/C9H13N2O9P.2Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,10,12,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1/i10+1,11+1;;

InChI key

KURVIXMFFSNONZ-SOIXSBQKSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service