Skip to Content
Merck

661201

Sigma-Aldrich

1-Z-3-Pyrrolidinone

96%

Synonym(s):

1-Carbobenzyloxy-3-pyrrolidinone, 1-Cbz-3-pyrrolidinone, 3-Oxo-1-pyrrolidinecarboxylic acid, phenylmethyl ester, 3-Oxopyrrolidine-1-carboxylic acid benzyl ester, N-Benzyloxycarbonyl-3-pyrrolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$92.20

$92.20


Availability

Available to ship onJanuary 06, 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$92.20

About This Item

Empirical Formula (Hill Notation):
C12H13NO3
CAS Number:
Molecular Weight:
219.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.20


Availability

Available to ship onJanuary 06, 2025Details


Request a Bulk Order

Quality Level

Assay

96%

impurities

(4% DCM)
4% DCM detected in first lot

refractive index

n20/D 1.5410

density

1.1897 g/mL at 25 °C

functional group

ketone
phenyl

SMILES string

O=C1CCN(C1)C(=O)OCc2ccccc2

InChI

1S/C12H13NO3/c14-11-6-7-13(8-11)12(15)16-9-10-4-2-1-3-5-10/h1-5H,6-9H2

InChI key

LMHWEUQNJRXMCD-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 3

This Item
654655721530
Quality Level

100

Quality Level

100

Quality Level

100

refractive index

n20/D 1.5410

refractive index

-

refractive index

-

density

1.1897 g/mL at 25 °C

density

-

density

-

impurities

(4% DCM)

impurities

-

impurities

-

functional group

ketone

functional group

hydroxyl, phenyl

functional group

ester

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service