Skip to Content
Merck
All Photos(1)

Key Documents

655953

Sigma-Aldrich

Triphosgene-13C3

99 atom % 13C

Synonym(s):

bis(trichloromethyl-13C) ester Carbonic-13C acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl313CO13COO13CCl3
CAS Number:
Molecular Weight:
299.73
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

bp

203-206 °C (lit.)

mp

79-83 °C (lit.)

mass shift

M+3

storage temp.

2-8°C

SMILES string

Cl[13C](Cl)(Cl)O[13C](=O)O[13C](Cl)(Cl)Cl

InChI

1S/C3Cl6O3/c4-2(5,6)11-1(10)12-3(7,8)9/i1+1,2+1,3+1

InChI key

UCPYLLCMEDAXFR-VMIGTVKRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service