Skip to Content
Merck
All Photos(1)

Key Documents

606251

Sigma-Aldrich

Phosgene-13C solution

~1 M in benzene, 99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13COCl2
CAS Number:
Molecular Weight:
99.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

mol wt

99.90 by atom % calculation

concentration

~1 M in benzene

mass shift

M+1

storage temp.

2-8°C

SMILES string

Cl[13C](Cl)=O

InChI

1S/CCl2O/c2-1(3)4/i1+1

InChI key

YGYAWVDWMABLBF-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Skin Corr. 1B - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service