Skip to Content
Merck
All Photos(1)

Key Documents

655538

Sigma-Aldrich

1,3-Bis(4-carboxyphenoxy)propane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16O6
CAS Number:
Molecular Weight:
316.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

310 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc(OCCCOc2ccc(cc2)C(O)=O)cc1

InChI

1S/C17H16O6/c18-16(19)12-2-6-14(7-3-12)22-10-1-11-23-15-8-4-13(5-9-15)17(20)21/h2-9H,1,10-11H2,(H,18,19)(H,20,21)

InChI key

VBISQLWPGDULSX-UHFFFAOYSA-N

Related Categories

Application

Building block for polyanhydrides, a class of bioerodible polymer with drug delivery applications.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moon Suk Kim et al.
Bio-medical materials and engineering, 15(3), 229-238 (2005-05-25)
p-Carboxyphenoxy propane (CPP) prepolymer consisting of 4 units and sebacic acid (SA) prepolymer consisting of about 10 units were synthesized by reacting CPP and SA in the presence of excess acetic anhydride, respectively. Polyanhydride, poly(CPP-SA) copolymers were copolymerized by a
A D'Emanuele et al.
Pharmaceutical research, 9(10), 1279-1283 (1992-10-01)
We report a study of the effects of polymer molecular weight on the erosion of polyanhydride copolymer matrices composed of 1,3-bis(p-carboxyphenoxy)-propane (CPP) and sebacic acid (SA) in aqueous solution. The erosion profile characteristically displays an induction period during which the
Sijian Hou et al.
Macromolecular bioscience, 7(5), 620-628 (2007-04-26)
To tailor the erosion rate of polyanhydrides while retaining their surface erosion characteristics, new three-component polyanhydrides of sebacic acid, 1,3-bis(p-carboxyphenoxy)propane and poly(ethylene glycol) were synthesized. The hydrophilicity of the polymer increased and its mechanical strength decreased with increasing PEG content.
Xiaohua Liu et al.
Biomaterials, 28(28), 4124-4131 (2007-06-20)
Intermittent (pulsatile) administration of parathyroid hormone (PTH) is known to improve bone micro-architecture, mineral density and strength. Therefore, daily injection of PTH has been clinically used for the treatment of osteoporosis. However, this regimen of administration is not convenient and
A J Domb et al.
Biomaterials, 15(9), 681-688 (1994-07-01)
The metabolic disposition and elimination process of the anhydride co-polymer poly[1,3-bis(p-carboxy-phenoxypropane):sebacic acid] 20:80 [P(CPP:Sa)20:80] implanted in the rat brain was studied. Two polymers were prepared, one with [14C]SA and unlabelled CPP, and the other co-polymer with [14C]CPP and unlabelled SA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service