Skip to Content
Merck
All Photos(2)

Key Documents

935646

Sigma-Aldrich

Ethylene sulfate

new
greener alternative

≥99%, anhydrous, battery grade

Synonym(s):

1,3,2-Dioxathiolane 2,2-dioxide, DTD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O4S
CAS Number:
Molecular Weight:
124.12
MDL number:
UNSPSC Code:
12352005

grade

anhydrous
battery grade

Quality Level

Assay

≥99%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤750 ppm (H2O)

bp

231 °C

mp

95-97 °C (lit.)

application(s)

battery manufacturing

greener alternative category

storage temp.

2-8°C

SMILES string

O=S1(=O)OCCO1

InChI

1S/C2H4O4S/c3-7(4)5-1-2-6-7/h1-2H2

InChI key

ZPFAVCIQZKRBGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene sulfate (DTD) is a polar cyclic sulfate ester. It is a white powder that is soluble in many of the polar organic solvents used in battery applications, including carbonates. Our battery grade ethylene sulfate is differentiated by its high purity, low trace water content, and low trace acid content, which avoid unwanted chemical reactions with the anode and increase capacity, safety, and life span in batteries

Application

Ethylene sulfate has a range of industrial applications. Our anhydrous battery grade ethylene sulfate is primarily used as an electrolyte additive to extend the lifetime of lithium-ion batteries and sodium-ion batteries.[,] Ethylene sulfate helps to improve the formation of a stable solid-electrolyte interphase (SEI) on the surface of the anode material, which can prevent reaction between the anode and the electrolyte, leading to better battery lifespan and more stable impedance across the interphase.[] DTD is often used in conjunction with other additives. One of the best-performing electrolyte formulations for graphite cells is a 1.0 M LiPF6 in a carbonate blend with 1% DTD and 2%VC.[,]

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Sulfate Electrolyte Additives on LiNi1/3Mn1/3Co1/3O2/Graphite Pouch Cell Lifetime: Correlation between XPS Surface Studies and Electrochemical Test Results
Madec L, et al.
The Journal of Physical Chemistry C, 118, 29608-29622 (2014)
Comparative study of 1,3-propane sultone, prop-1-ene-1,3-sultone and ethylene sulfate as film-forming additives for sodium ion batteries
Zhang Q, et al.
Journal of Power Sources, 541, 231726-231726 (2022)
A Wide Range of Testing Results on an Excellent Lithium-Ion Cell Chemistry to be used as Benchmarks for New Battery Technologies
Jessie E, et al.
Journal of the Electrochemical Society, 166, A3031-A3031 (2019)
Ethylene sulfate as film formation additive to improve the compatibility of graphite electrode for lithium-ion battery
Xuecheng L, et al.
Ionics, 20, 795-801 (2014)
Ethylene sulfate as film formation additive to improve the compatibility of graphite electrode for lithium-ion battery
Xuecheng L, et al.
Ionics, 20, 795-801 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service