Skip to Content
Merck
All Photos(1)

Key Documents

651486

Sigma-Aldrich

Gallium arsenide

(single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm

Synonym(s):

Gallium monoarsenide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaAs
CAS Number:
Molecular Weight:
144.64
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

(single crystal substrate)

resistivity

≥1E7 Ω-cm

diam. × thickness

2 in. × 0.5 mm

density

5.31 g/mL at 25 °C (lit.)

semiconductor properties

<100>

SMILES string

[Ga]#[As]

InChI

1S/As.Ga

InChI key

JBRZTFJDHDCESZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

Mobility >=4500 cm2 · V-1 · S-1
Undoped (Si-type semiconductor), EPD < 5 × 104 cm-2, growth technique = LEC & HB

Physical form

cubic (a = 5.6533 Å)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 1B - STOT RE 1

Target Organs

Respiratory system,hematopoietic system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anand Kumar Tatikonda et al.
Biosensors & bioelectronics, 45, 201-205 (2013-03-19)
Microelectronic-based sensors are ideal for real-time continuous monitoring of health states due to their low cost of production, small size, portability, and ease of integration into electronic systems. However, typically semiconductor-based devices cannot be operated in aqueous solutions, especially in
V V Solovyev et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(2), 025801-025801 (2012-11-28)
Temperature-dependent reflectivity and photoluminescence spectra are studied for undoped ultra-wide 150 and 250 nm GaAs quantum wells. It is shown that spectral features previously attributed to a size quantization of the exciton motion in the z-direction coincide well with energies
I I Yakimenko et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 072201-072201 (2013-01-19)
We analyze the occurrence of local magnetization and the effects of electron localization in different models of quantum point contacts (QPCs) using spin-relaxed density functional theory (DFT/LSDA) by means of numerical simulations. In the case of soft confinement potentials the
Andrew Lee et al.
Optics express, 20(20), 22181-22187 (2012-10-06)
We report the first room-temperature continuous-wave operation of III-V quantum-dot laser diodes monolithically grown on a Si substrate. Long-wavelength InAs/GaAs quantum-dot structures were fabricated on Ge-on-Si substrates. Room-temperature lasing at a wavelength of 1.28 μm has been achieved with threshold
Eli Fahrenkrug et al.
Journal of the American Chemical Society, 135(1), 330-339 (2012-12-26)
Crystalline GaAs (c-GaAs) has been prepared directly through electroreduction of As(2)O(3) dissolved in an alkaline aqueous solution at a liquid gallium (Ga(l)) electrode at modest temperatures (T ≥ 80 °C). Ga(l) pool electrodes yielded consistent electrochemical behavior, affording repetitive measurements

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service