Skip to Content
Merck
All Photos(1)

Key Documents

648159

Sigma-Aldrich

PAMAM dendrimer, cystamine core, generation 5, solution

10 wt. % in methanol

Synonym(s):

PAMAM dendrimer, cystamine core, generation 5.0 solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2CH2CH2SSCH2CH2NH2]:(G=5);dendri PAMAM(NH2)128
Molecular Weight:
28917.00
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amino surface groups

form

solution

mol wt

generation 5

feature

core type cystamine core

concentration

10 wt. % in methanol

no. Surface Groups

128

bp

64.7 °C

density

0.860 g/mL at 25 °C

storage temp.

−20°C

Packaging

unit weight includes solvent

Legal Information

For research use only

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang, B. H., Tomalia, D. A.
Journal of Luminescence, 111, 215-223 (2005)
DeMattei, C. R., Huang, B. H., Tomalia, D. A.
Nano Letters, 4, 771-777 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service