Skip to Content
Merck
All Photos(1)

Key Documents

412392

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 1.5 solution

20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=1.5);dendri PAMAM(NHCH2CH2COONa)16
CAS Number:
Molecular Weight:
2934.56
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

sodium carboxylate surface groups

Quality Level

form

liquid

mol wt

generation 1.5

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

16

refractive index

n20/D 1.365

bp

65 °C

density

0.86 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CCN(CCNC(=O)CCN(CCNC(=O)CCN(CCN(CCC(=O)NCCN(CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC([O-])=O

InChI

1S/C110H192N26O44.16Na/c137-83(115-33-73-127(53-13-95(149)150)54-14-96(151)152)1-41-123(42-2-84(138)116-34-74-128(55-15-97(153)154)56-16-98(155)156)69-29-111-91(145)9-49-135(50-10-92(146)112-30-70-124(43-3-85(139)117-35-75-129(57-17-99(157)158)58-18-100(159)160)44-4-86(140)118-36-76-130(59-19-101(161)162)60-20-102(163)164)81-82-136(51-11-93(147)113-31-71-125(45-5-87(141)119-37-77-131(61-21-103(165)166)62-22-104(167)168)46-6-88(142)120-38-78-132(63-23-105(169)170)64-24-106(171)172)52-12-94(148)114-32-72-126(47-7-89(143)121-39-79-133(65-25-107(173)174)66-26-108(175)176)48-8-90(144)122-40-80-134(67-27-109(177)178)68-28-110(179)180;;;;;;;;;;;;;;;;/h1-82H2,(H,111,145)(H,112,146)(H,113,147)(H,114,148)(H,115,137)(H,116,138)(H,117,139)(H,118,140)(H,119,141)(H,120,142)(H,121,143)(H,122,144)(H,149,150)(H,151,152)(H,153,154)(H,155,156)(H,157,158)(H,159,160)(H,161,162)(H,163,164)(H,165,166)(H,167,168)(H,169,170)(H,171,172)(H,173,174)(H,175,176)(H,177,178)(H,179,180);;;;;;;;;;;;;;;;/q;16*+1/p-16

InChI key

KKDVRAVJOOYSGF-UHFFFAOYSA-A

Physical form

Contains 16 surface carboxylate groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service