648116
Methyl 3-(bromomethyl)benzoate
97%
Synonym(s):
3-(Bromomethyl)benzoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
97%
form
solid
bp
112-114 °C/3 mmHg (lit.)
mp
41-45 °C (lit.)
functional group
bromo
ester
storage temp.
2-8°C
SMILES string
COC(=O)c1cccc(CBr)c1
InChI
1S/C9H9BrO2/c1-12-9(11)8-4-2-3-7(5-8)6-10/h2-5H,6H2,1H3
InChI key
YUHSMQQNPRLEEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service