Skip to Content
Merck
Methyl 3-(bromomethyl)benzoate 97%

Methyl 3-(bromomethyl)benzoate

Methyl 3-(bromomethyl)benzoate 97%
Synonym(s):
3-(Bromomethyl)benzoic acid methyl ester
Empirical Formula (Hill Notation):
C9H9BrO2
CAS Number:
Molecular Weight:
229.07

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing