All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H9NO
CAS Number:
Molecular Weight:
135.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
solid
bp
118-120 °C/20 mmHg (lit.)
mp
55-60 °C (lit.)
density
1.11 g/mL at 25 °C (lit.)
functional group
amine
oxime
phenyl
storage temp.
−20°C
SMILES string
C\C(=N/O)c1ccccc1
InChI
1S/C8H9NO/c1-7(9-10)8-5-3-2-4-6-8/h2-6,10H,1H3/b9-7+
InChI key
JHNRZXQVBKRYKN-VQHVLOKHSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tomasz Kosmalski et al.
ChemistryOpen, 7(7), 551-557 (2018-08-02)
In this study, we present a convenient method for the synthesis of oxime ethers by reacting oximes with various chlorides (alkyl, functionalized alkyl, and benzyl) and with the subsequent use of a super base-pulverized potassium hydroxide in DMSO. The reactions
Rajkumar Kore et al.
Dalton transactions (Cambridge, England : 2003), 47(23), 7795-7803 (2018-06-01)
Two solids of differing Lewis acidities, triethylammonium tetrachlorozincate ([HN222]2[ZnCl4]) and AlCl3, have been combined across 0.1 to 0.9 mole fractions (with respect to [HN222]2[ZnCl4]), and homogeneous solid double salts or mixed metal [HN222]2x[(1 - x)AlCl3 + xZnCl4] double salt ionic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service