Skip to Content
Merck
All Photos(2)

Key Documents

332828

Sigma-Aldrich

2-Butanone oxime

99%

Synonym(s):

Methyl ethyl ketoxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(=NOH)CH3
CAS Number:
Molecular Weight:
87.12
Beilstein:
1698241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

vapor pressure

<8 mmHg ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

59-60 °C/15 mmHg (lit.)

solubility

water: soluble 100 g/L at 25 °C

density

0.924 g/mL at 25 °C (lit.)

functional group

amine
oxime

SMILES string

CC\C(C)=N\O

InChI

1S/C4H9NO/c1-3-4(2)5-6/h6H,3H2,1-2H3/b5-4+

InChI key

WHIVNJATOVLWBW-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Butanone oxime is an antioxidant and blocking agent used in the synthesis of water-dispersible blocked polyurethane.

Application

2-Butanone oxime was employed as reagent and solvent in the syntheses of ketoimine and 2,4-dipyridyl-1,3,5-triazapentadiene palladium(II) complexes. It was also used in the synthesis of novel acetaldiimine cobalt complex, [CoI2{((CH3CH2)(CH3)C=NO)2C(CH3)2}].

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Blood, Central nervous system, Upper respiratory tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.5 °F - closed cup

Flash Point(C)

61.97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul E Newton et al.
Inhalation toxicology, 14(12), 1249-1260 (2002-11-28)
Methylethylketoxime, also known as MEKO or 2-butanone oxime (CAS No. 96-29-7), is a clear, colorless to light yellow liquid at room temperature. It is an industrial antioxidant used as an antiskinning agent in alkyd paint, an industrial blocking agent for
Michael J Derelanko et al.
Drug and chemical toxicology, 31(1), 97-114 (2007-12-29)
Acute and repeated oral and dermal rat toxicology studies of standard designs were conducted on four methyl ethyl ketoxime (MEKO) silanes and four methyl isobutyl ketoxime (MIBKO) silanes. Each compound contained either MEKO or MIBKO groups (but not both) and
Developmental toxicity studies of methyl ethyl ketoxime (MEKO) in rats and rabbits
Michael D J et al.
Drug and Chemical Toxicology, 26, 147-168 (2003)
Maximilian N Kopylovich et al.
Dalton transactions (Cambridge, England : 2003), (16)(16), 3074-3084 (2009-04-09)
[2 + 3] cycloaddition of the pyrrolin N-oxide (-)O[upper bond 1 start](+)N[double bond, length as m-dash]CHCH(2)CH(2)C[upper bond 1 end]Me(2) with 2-cyanopyridine NC(5)H(4)(2-CN) in the presence of PdCl(2) at room temperature in acetone gives the ketoimine palladium(II) complex cis-[PdCl(2){N(=[upper bond 1
P E Newton et al.
Inhalation toxicology, 13(12), 1093-1116 (2001-11-07)
To evaluate the oncogenic potential of methylethylketoxime (MEKO), CD-1 mice (50/sex/group) and F-344 rats (50/sex/group) were coexposed 6 h/day, 5 days/wk for 18 mo (mice) or 26 mo (rats) via whole-body inhalation exposures to target vapor concentrations of 0, 15

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service