Skip to Content
Merck
All Photos(1)

Key Documents

646369

Sigma-Aldrich

6,6-Dimethylfulvene

technical grade

Synonym(s):

5-(1-Methylethylidene)-1,3-cyclopentadiene, 5-Isopropylidene-1,3-cyclopentadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
Beilstein:
1616308
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

refractive index

n20/D 1.548 (lit.)

bp

76-77 °C/50 mmHg (lit.)

density

0.881 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C(C)=C1/C=CC=C1

InChI

1S/C8H10/c1-7(2)8-5-3-4-6-8/h3-6H,1-2H3

InChI key

WXACXMWYHXOSIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service