Skip to Content
Merck
All Photos(1)

Key Documents

64410

Sigma-Aldrich

DL-Methionine sulfone

≥99.0% (NT)

Synonym(s):

DL-2-Amino-4-(methylsulfonyl)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2CH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
181.21
Beilstein:
2089918
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (NT)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

~250 °C (dec.)

application(s)

peptide synthesis

SMILES string

CS(=O)(=O)CCC(N)C(O)=O

InChI

1S/C5H11NO4S/c1-11(9,10)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

UCUNFLYVYCGDHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae Hee Han et al.
Asian-Australasian journal of animal sciences, 30(8), 1150-1159 (2017-02-12)
This study was conducted to evaluate various wheat supplementation levels on growth performance, blood profiles, nutrient digestibility, and pork quality in growing-finishing pigs. A total of 120 growing pigs ([Yorkshire×Landrace]×Duroc), with an average 27.75± 1.319 kg body weight, were used
Vincenza M Coiro et al.
Protein science : a publication of the Protein Society, 13(11), 2979-2991 (2004-10-23)
Glutamate synthase (GltS) is a complex iron-sulfur flavoprotein that catalyzes the reductive transfer of L-glutamine amide group to the C2 carbon of 2-oxoglutarate yielding two molecules of L-glutamate. Molecular dynamics calculations in explicit solvent were carried out to gain insight
C Binda et al.
Structure (London, England : 1993), 8(12), 1299-1308 (2001-02-24)
The complex iron-sulfur flavoprotein glutamate synthase catalyses the reductive synthesis of L-glutamate from 2-oxoglutarate and L-glutamine, a reaction in the plant and bacterial pathway for ammonia assimilation. The enzyme functions through three distinct active centers carrying out L-glutamine hydrolysis, conversion
C M Parsons et al.
The British journal of nutrition, 46(1), 77-86 (1981-07-01)
1. The relative potencies of three lysine, one tryptophan and six methionine analogues to their corresponding l-amino acids were determined. 2. Male poults, 9 d of age, were used in five 14 d experiments. Experimental diets were formed by adding
F J Cejudo et al.
Biochemical and biophysical research communications, 123(2), 431-437 (1984-09-17)
Addition of NH4Cl at low concentrations to Azotobacter chroococcum cells caused an immediate cessation of nitrogenase activity, which was recovered once the added NH+4 was exhausted from the medium. In the presence of inhibitors of ammonium assimilation, such as L-methionine-DL-sulfoximine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service