Skip to Content
Merck
All Photos(1)

Key Documents

N1398

Sigma-Aldrich

DL-Norleucine

≥98% (TLC)

Synonym(s):

(±)-2-Aminocaproic acid, 2-Aminohexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(NH2)COOH
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721748
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

DL-Norleucine,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white to off-white

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCCCC(N)C(O)=O

InChI

1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)

InChI key

LRQKBLKVPFOOQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DL-Norleucine is a nonsulfur analogue of methionine that stimulates synthesis of cephalosporin C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measuring and Modeling Activity Coefficients in Aqueous Amino-Acid Solutions
Held, C, Cameretti, L, and Sadowski, G.
Ind. and Eng. Chem., 50, 131-141 (2011)
Glenn S Murphy et al.
Anesthesiology, 122(5), 1112-1122 (2015-04-04)
The intensity of pain after cardiac surgery is often underestimated, and inadequate pain control may be associated with poorer quality of recovery. The aim of this investigation was to examine the effect of intraoperative methadone on postoperative analgesic requirements, pain
Dharmesh R Chejara et al.
Carbohydrate polymers, 135, 324-333 (2015-10-11)
In this article a novel bio-injectable algin-aminocaproic acid (Alg-ACA) tri-stimuli responsive thixogel system is reported. The designed soft thixotrophic hydrogel (thixogel) was characterized using various analytical techniques such as FT-IR, NMR, SEM, AFM and DSC. The soft thixogel system was
Raj Shah et al.
Cancer research, 79(8), 1799-1809 (2019-04-17)
Aberrant glutamatergic signaling has been implicated in altered metabolic activity in many cancer types, including malignant melanoma. Previously, we have illustrated the role of metabotropic glutamate receptor 1 (GRM1) in neoplastic transformation of melanocytes in vitro and spontaneous metastatic melanoma
Nabaneeta Mukhopadhyay et al.
Physical chemistry chemical physics : PCCP, 17(6), 4293-4310 (2015-01-13)
Blends of polypropylene (PP) and polyamide 6 (PA6) with multiwalled carbon nanotubes (MWNTs) were prepared using different processing strategies in a twin-screw micro-compounder. The effect of MWNTs on the crystallization behaviour of the PP phase and the PA6 phase of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service