Recommended Products
Assay
96%
refractive index
n20/D 1.4900
bp
159-160 °C/760 mmHg (lit.)
density
0.946 g/mL at 25 °C
functional group
amine
thioether
SMILES string
CCSCCN
InChI
1S/C4H11NS/c1-2-6-4-3-5/h2-5H2,1H3
InChI key
HJCTVUWPHAZTLI-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
134.6 °F - closed cup
Flash Point(C)
57 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service