Skip to Content
Merck
All Photos(1)

Key Documents

C121509

Sigma-Aldrich

Cystamine dihydrochloride

96%

Synonym(s):

2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SSCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
225.20
Beilstein:
3616850
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

powder

mp

217-220 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].NCCSSCCN

InChI

1S/C4H12N2S2.2ClH/c5-1-3-7-8-4-2-6;;/h1-6H2;2*1H

InChI key

YUFRRMZSSPQMOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cystamine dihydrochloride can be utilized as a building block in the synthesis of disulfide cross-linked oligodeoxyribonucleotides and psammaplin A . It can be also used to functionalize PGMA (poly(glycidyl methacrylate) microsphere) by introducing sulfhydryl groups for the further fabrication of silver nanoparticles (AgNPs).
Heparin antagonist and sulfhydryl modifying reagent

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ synthesis of monodisperse silver nanoparticles on sulfhydryl-functionalized poly (glycidyl methacrylate) microspheres for catalytic reduction of 4-nitrophenol
Zhang W, et al.
Industrial & Engineering Chemistry Research, 54(25), 6480-6488 (2015)
Synthesis and characterization of disulfide cross-linked oligonucleotides
Ferentz A E, et al.
Journal of the American Chemical Society, 115(20), 9006-9014 (1993)
Qianjiao Yang et al.
Molecules (Basel, Switzerland), 15(12), 8784-8795 (2010-12-04)
Psammaplin F, an unsymmetrical disulfide bromotyrosine, was isolated from the sponge Pseudoceratina purpurea in 2003. We reported here the first total synthesis of psammaplin F in 12% overall yield by employing Cleland's reagent reduction as key step. The longest linear
Zhongshan Liu et al.
Journal of chromatography. A, 1342, 70-77 (2014-04-15)
A polyhedral oligomeric silsesquioxane (POSS) hybrid monolith was simply prepared by using octaglycidyldimethylsilyl POSS (POSS-epoxy) and cystamine dihydrochloride as monomers via ring-opening polymerization. The effects of composition of prepolymerization solution and polycondensation temperature on the morphology and permeability of monolithic
Thorsten Overath et al.
Molecular & cellular proteomics : MCP, 11(8), 467-477 (2012-05-05)
The post-translational modification of proteins with O-GlcNAc is involved in various cellular processes including signal transduction, transcription, translation, and nuclear transport. This transient protein modification enables cells or tissues to adapt to nutrient conditions or stress. O-Glycosylation of the 26

Articles

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service