637947
2-Bromoethanol-2-13C
99 atom % 13C
Synonym(s):
Ethylene-2-13C bromohydrin
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
bp
56-57 °C/20 mmHg (lit.)
density
1.777 g/mL at 25 °C (lit.)
mass shift
M+1
SMILES string
OC[13CH2]Br
InChI
1S/C2H5BrO/c3-1-2-4/h4H,1-2H2/i1+1
InChI key
LDLCZOVUSADOIV-OUBTZVSYSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service