All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C4H5ClN4
CAS Number:
Molecular Weight:
144.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
137-141 °C (lit.)
functional group
chloro
hydrazine
SMILES string
NNc1ccc(Cl)nn1
InChI
1S/C4H5ClN4/c5-3-1-2-4(7-6)9-8-3/h1-2H,6H2,(H,7,9)
InChI key
FXYQRYGWWZKUFV-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Lindvall et al.
Chemico-biological interactions, 94(2), 83-99 (1995-02-01)
(Z)-3-(4-Bromophenyl)-3-(3-pyridyl)allylamine (CPP 200) is transformed to the corresponding chloroimine by hypochlorite ion (ClO-) formed in the presence of myeloperoxidase. A scheme for this transformation is given. The influence of various compounds on this process has been studied. Cysteamine, cysteine and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service