616850
1,1,4,4-Tetraphenyl-1,3-butadiene-d22
98 atom % D
About This Item
Recommended Products
isotopic purity
98 atom % D
form
solid
mp
207-209 °C
mass shift
M+22
SMILES string
[2H]c1c([2H])c([2H])c(c([2H])c1[2H])\C(=C([2H])\C([2H])=C(\c2c([2H])c([2H])c([2H])c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H])c4c([2H])c([2H])c([2H])c([2H])c4[2H]
InChI
1S/C28H22/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26/h1-22H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D,15D,16D,17D,18D,19D,20D,21D,22D
InChI key
KLCLIOISYBHYDZ-SMJVRUDNSA-N
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service