Skip to Content
Merck
All Photos(3)

Key Documents

434388

Sigma-Aldrich

Toluene-d8

99.6 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD3
CAS Number:
Molecular Weight:
100.19
Beilstein:
1909300
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.6 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤100 ppm water

refractive index

n20/D 1.494 (lit.)

bp

110 °C (lit.)

mp

-84 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

mass shift

M+8

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])[2H]

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D

InChI key

YXFVVABEGXRONW-JGUCLWPXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene-d8 is a deuterated NMR solvent useful in NMR-based research and analyses. Spin–lattice relaxation time (T 1), viscosity and density have been measured over -35 to 200°C range. The temperature dependent chemical shifts in the proton NMR spectra of α,,β-unsaturated ketones in toluene-d8 has been studied.

Application


  • Improved Miniaturized Linear Ion Trap Mass Spectrometer: Utilizes Toluene-d₈ in lithographically patterned plates to enhance the ejection efficiency and resolution of mass spectrometry. This advancement in analytical technology significantly improves the detection of complex molecular structures, crucial for pharmaceutical and biochemical research (Tian Y et al., 2018).

  • Structure-function relationships in high-density docosylsilane bonded stationary phases: Employs Toluene-d₈ to elucidate the effects of common solvents on stationary phase structures by Raman spectroscopy. This research enhances understanding of chromatographic behaviors, influencing the development of more effective and robust chromatographic systems for environmental and material science applications (Liao Z et al., 2008).

  • Characterization of Pore Structure in a Nanoporous Low-dielectric-constant Thin Film: Toluene-d₈ is used in neutron and X-ray porosimetry to characterize the porous structure of innovative low-dielectric materials. This application is critical in the development of advanced electronic materials, promoting enhanced performance and miniaturization of electronic devices (Hedden RC et al., 2004).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solvent Effects in Nuclear Magnetic Resonance Spectroscopy. IX. 1 A Variable-Temperature Study of α,β-Unsaturated Ketones in Toluene-d8 Solution.
Ronayne J, et al.
Journal of the American Chemical Society, 88(22), 5288-5292 (1966)
Fourier transform NMR in liquids at high pressure. III. Spin-lattice relaxation in toluene-d8.
Wilbur DJ and Jonas J.
J. Chem. Phys. , 62(7), 2800-2807 (1975)
Freddy H Havaldar et al.
Journal of labelled compounds & radiopharmaceuticals, 58(4), 163-165 (2015-02-19)
A d4 -labeled isotopomer of deferasirox was synthesized as internal standard for use in a LC/mass spectroscopy (MS)/MS method developed for the simultaneous quantitative determination of deferasirox in human serum. d4 -deferasirox was synthesized from d8 -toluene.
Xiaoyong Lu et al.
Molecules (Basel, Switzerland), 19(9), 14292-14303 (2014-09-13)
Gated molecular encapsulations, with baskets of type 1, are postulated to occur by the mechanism in which solvent molecule penetrates the inner space of 1, through one of its apertures, while the residing guest simultaneously departs the cavity. In the
Didier G Diomande et al.
Analytical chemistry, 87(13), 6600-6606 (2015-06-13)
The natural xanthines caffeine, theobromine, and theophylline are of major commercial importance as flavor constituents in coffee, cocoa, tea, and a number of other beverages. However, their exploitation for authenticity, a requirement in these commodities that have a large origin-based

Articles

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service