Skip to Content
Merck
All Photos(1)

Key Documents

614106

Sigma-Aldrich

Cholic acid-2,2,3,4,4-d5

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D5H35O5
CAS Number:
Molecular Weight:
413.60
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

200-201 °C (lit.)

mass shift

M+5

SMILES string

[2H][C@@]1(O)C([2H])([2H])C[C@@]2(C)[C@@](C[C@@H](O)[C@]3([H])[C@]2([H])C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](CCC(O)=O)C)([H])C1([2H])[2H]

InChI

1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1/i8D2,10D2,15D

InChI key

BHQCQFFYRZLCQQ-VRLVFLRJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Gabbia et al.
Nutrients, 11(7) (2019-07-18)
Since nutrition might have a significant impact on liver function, we analyzed the early effect of Western-type diet on hepatic tissue and lipid and drug metabolism in Wistar-Kyoto rats (n = 8); eight rats fed with a standard diet were
Suocheng Hui et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(6), 8558-8573 (2020-05-03)
Mounting evidence has linked dietary capsaicin (CAP) consumption to the improvement of glucose homeostasis; however, the underlying mechanisms still need to be further elucidated. Male mice were fed a high-fat diet (HFD) with CAP administration for 8 weeks, gut microbiota, bile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service