Skip to Content
Merck
All Photos(1)

Key Documents

694940

Sigma-Aldrich

Linolenic Acid-1-13C

99 atom % 13C, 95% (CP)

Synonym(s):

Linolenic Acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)3(CH2)713CO2H
Molecular Weight:
279.42
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

95% (CP)

form

liquid

bp

230-232 °C/1 mmHg

mp

-11 °C

density

0.917 g/mL at 25 °C

mass shift

M+1

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/CCCCCCC[13C](O)=O

InChI

1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-/i18+1

InChI key

DTOSIQBPPRVQHS-JAYTYYOISA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P G Toral et al.
Journal of dairy science, 102(2), 1213-1223 (2018-12-26)
The basis for marine lipid-induced milk fat depression (MFD) has not been established yet, but recent reports suggest the putative contribution of shifts in the ruminal metabolism of α-linolenic acid (ALA). To test this hypothesis, an isotopic tracer approach was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service