609803
Methanol-17O
20 atom % 17O
Synonym(s):
Methyl alcohol-17O
About This Item
Recommended Products
isotopic purity
20 atom % 17O
Assay
99% (CP)
mol wt
32.25 by atom % calculation (20 atom % 17O)
bp
65.4 °C (lit.)
density
0.816 g/mL at 25 °C
mass shift
M+1
SMILES string
C[17OH]
InChI
1S/CH4O/c1-2/h2H,1H3/i2+1
InChI key
OKKJLVBELUTLKV-VQEHIDDOSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service