Skip to Content
Merck
All Photos(2)

Key Documents

244600

Sigma-Aldrich

Cadmium selenide

-325 Mesh particle size, 99.99% trace metals basis, electronic grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdSe
CAS Number:
Molecular Weight:
191.37
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

electronic grade

Quality Level

Assay

99.99% trace metals basis

form

powder

density

5.81 g/mL at 25 °C (lit.)

SMILES string

[Se]=[Cd]

InChI

1S/Cd.Se

InChI key

AQCDIIAORKRFCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

CdSe crystal can exist in solid hexagonal or cubic structure. CdSe behaves as an n-type semiconductor, with a bandgap of 1.74 eV (300K).1

Application

Dispersions with CdS (Aldrich products 208183, 217921) deposited on glass or Si substrates results in tunable red, yellow, or green fluorescent material.[1]

Packaging

Packaged in glass bottles

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney,Bone

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Wang et al.
Journal of the American Chemical Society, 125(10), 2830-2831 (2003-03-06)
Indiscriminate adsorption of nanoparticles (NPs) significantly complicates the preparation of mesoscale NP patterns considered as enabling technology for many devices and processes. Instead of selected chemical functionalization of the substrate surface prior to the assembly of nanocolloids, the required optical
J W Fellowes et al.
Nanotechnology, 24(14), 145603-145603 (2013-03-20)
Luminescent quantum dots were synthesized using bacterially derived selenide (Se(II-)) as the precursor. Biogenic Se(II-) was produced by the reduction of Se(IV) by Veillonella atypica and compared directly against borohydride-reduced Se(IV) for the production of glutathione-stabilized CdSe and β-mercaptoethanol-stabilized ZnSe
Lu Kong et al.
Journal of nanoscience and nanotechnology, 12(11), 8258-8265 (2013-02-21)
Quantum dots are being widely used in physics and in the biomedical industry in recent years due to their excellent optical characteristics. However, studies have shown that cadmium selenide core-shell quantum dots exhibit cytotoxicity. The present study investigates the induction
David T R Stewart et al.
Environmental toxicology and chemistry, 32(6), 1288-1294 (2013-02-19)
The present study addresses the bioaccumulation behavior of cadmium selenide quantum dots by Eisenia andrei earthworms in a terrestrial environment. Earthworms were exposed to quantum dot-treated soil for up to 4 wk and analyzed for cadmium and selenium concentration using
Yong Li et al.
ACS nano, 7(3), 2240-2248 (2013-02-13)
Microbial cells have shown a great potential to biosynthesize inorganic nanoparticles within their orderly regulated intracellular environment. However, very little is known about the mechanism of nanoparticle biosynthesis. Therefore, it is difficult to control intracellular synthesis through the manipulation of

Articles

Review the potential of self-assembled multilayer gate dielectric films fabricated from silane precursors for organic, inorganic, and transparent TFT and for TFT circuitry and OLED displays.

Review the potential of self-assembled multilayer gate dielectric films fabricated from silane precursors for organic, inorganic, and transparent TFT and for TFT circuitry and OLED displays.

Review the potential of self-assembled multilayer gate dielectric films fabricated from silane precursors for organic, inorganic, and transparent TFT and for TFT circuitry and OLED displays.

Review the potential of self-assembled multilayer gate dielectric films fabricated from silane precursors for organic, inorganic, and transparent TFT and for TFT circuitry and OLED displays.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service