Skip to Content
Merck
All Photos(1)

Key Documents

609781

Sigma-Aldrich

Phosphoric acid-16O4 solution

70 wt. % in D2O, 99.9 atom % 16O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3P16O4
Molecular Weight:
97.98
UNSPSC Code:
12352301
PubChem Substance ID:

description

18O-depleted

isotopic purity

99.9 atom % 16O

concentration

70 wt. % in D2O

mass shift

depleted

storage temp.

−20°C

SMILES string

[16OH]P([16OH])([16OH])=[16O]

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)/i1+0,2+0,3+0,4+0

InChI key

NBIIXXVUZAFLBC-KTAHLDKPSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit Kumar Sinha et al.
PloS one, 9(4), e95319-e95319 (2014-04-18)
Oxidative stress and the antioxidant response induced by high environmental ammonia (HEA) were investigated in the liver and gills of three freshwater teleosts differing in their sensitivities to ammonia. The highly ammonia-sensitive salmonid Oncorhynchus mykiss (rainbow trout), the less ammonia
Jesús Torres et al.
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft, 200, 126-133 (2015-05-02)
The aim of this study was to test bone tissue response to monetite granules in comparison with intramembranous autologous bone graft in a rabbit calvaria critical size defect model. Novel monetite granules were synthesized by thermal conversion of set brushite
Manabu Ueno et al.
Nature communications, 6, 6332-6332 (2015-03-11)
Alveolar macrophages play a crucial role in the pathogenesis of emphysema, for which there is currently no effective treatment. Bisphosphonates are widely used to treat osteoclast-mediated bone diseases. Here we show that delivery of the nitrogen-containing bisphosphonate alendronate via aerosol
Kangmin Chon et al.
Water research, 81, 388-397 (2015-07-05)
In this study, the changes in UV absorbance at 254 nm (UVA254) and electron donating capacity (EDC) were investigated as surrogate indicators for assessing removal of micropollutants and bromate formation during ozonation of wastewater effluents. To measure the EDC, a novel
Mian Qi et al.
Journal of the American Chemical Society, 136(43), 15366-15378 (2014-10-18)
Distance measurement in the nanometer range by electron paramagnetic resonance spectroscopy (EPR) in combination with site-directed spin labeling is a very powerful tool to monitor the structure and dynamics of biomacromolecules in their natural environment. However, in-cell application is hampered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service