176753
Phosphoric acid-d3 solution
85 wt. % in D2O, 98 atom % D
Synonym(s):
Trideuterophosphoric acid
About This Item
Recommended Products
isotopic purity
98 atom % D
Quality Level
concentration
85 wt. % in D2O
technique(s)
NMR: suitable
refractive index
n20/D 1.427 (lit.)
bp
158 °C/760 mmHg (lit.)
mp
41-44 °C (lit.)
density
1.736 g/mL at 25 °C
mass shift
M+3
SMILES string
[2H]OP(=O)(O[2H])O[2H]
InChI
1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)/i/hD3
InChI key
NBIIXXVUZAFLBC-ZRLBSURWSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service